Mouse recombinant Thymic stromal lymphopoietin
Fournisseur: ProSci Inc.
92-467
PRSI92-467EA 588 EUR InStock
PRSI92-467
Mouse recombinant Thymic stromal lymphopoietin
Protéines et Peptides
Thymic stromal lymphopoietin (TSLP) is a protein belonging to the cytokine family, contains 140 amino acids. It is known to play an important role in the maturation of T cell populations through activation of antigen presenting cells. TSLP induces the release of T-cell-attracting chemokines from monocytes and, in particular, enhances the maturation of CD11c+ dendritic cells. It can induce allergic inflammation by directly activating mast cells. TSLP is produced mainly by non-hematopoietic cells such as fibroblasts, epithelial cells and different types of stromal or stromal-like cells. These cells are located in regions where TSLP activity is required.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité