Mouse recombinant SEMA4C
Fournisseur: ProSci Inc.
92-468
PRSI92-468EA 852 EUR InStock
PRSI92-468
Mouse recombinant SEMA4C
Protéines et Peptides
Semaphorin-4C is a protein which belongs to the semaphorin family, contains 1 Ig-like C2-type domain, 1 PSI domain, 1 Sema domain. As cell surface receptor for PLXNB2, it plays an important role in cell-cell signalling. PLXNB2 binding promotes downstream activation of RHOA and phosphorylation of ERBB2 at 'Tyr-1248'. It required for normal brain development, axon guidance and cell migration, Probable signalling receptor which may play a role in myogenic differentiation through activation of the stress-activated MAPK cascade.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité