Human recombinant Ephrin type B Receptor 2 (from cells)
Fournisseur: ProSci Inc.
92-471
PRSI92-471EA 588 EUR InStock
PRSI92-471
Human recombinant Ephrin type B Receptor 2 (from cells)
Protéines et Peptides
Ephrin type-B receptor 2(EPHB2) belongs to the protein kinase superfamily and Ephrin receptor subfamily. EPHB2 contains 1 Eph LBD domain, 2 fibronectin type-III domains, 1 protein kinase domain and 1 SAM domain. Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité