Human recombinant Ephrin A5 (from cells)
Fournisseur: ProSci Inc.
92-474
PRSI92-474EA 654 EUR InStock
PRSI92-474
Human recombinant Ephrin A5 (from cells)
Protéines et Peptides
Ephrin-A5 (EFNA5) belongs to the ephrin family,contains 1 ephrin RBD (ephrin receptor-binding) domain. Ephrin-A5 is a cell surface GPI-bound ligand for Eph receptors, a family of receptor tyrosine kinases which are crucial for migration, repulsion and adhesion during neuronal, vascular and epithelial development. It binds promiscuously Eph receptors residing on adjacent cells, leading to contact-dependent bidirectional signalling into neighboring cells. The interaction of EFNA5 with EPHA5 also mediates communication between pancreatic islet cells to regulate glucose-stimulated insulin secretion. Cognate/functional ligand for EPHA7, their interaction regulates brain development modulating cell-cell adhesion and repulsion.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité