Human recombinant IL-2 R alpha (from cells)
Fournisseur: ProSci Inc.
92-475
PRSI92-475EA 654 EUR InStock
PRSI92-475
Human recombinant IL-2 R alpha (from cells)
Protéines et Peptides
Interleukin-2 receptor subunit alpha (IL2RA) is a single-pass type I membrane protein, contains 2 Sushi (CCP/SCR) domains. The interleukin 2 (IL2) receptor alpha (IL2RA) and beta (IL2RB) chains, together with the common gamma chain (IL2RG), constitute the high-affinity IL2 receptor. Homodimeric alpha chains (IL2RA) result in low-affinity receptor, while homodimeric beta (IL2RB) chains produce a medium-affinity receptor. Normally an integral-membrane protein, soluble IL2RA has been isolated and determined to result from extracellular proteolyisis.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité