Mouse recombinant HVEM
Fournisseur: ProSci Inc.
92-488
PRSI92-488EA 343 EUR InStock
PRSI92-488
Mouse recombinant HVEM
Protéines et Peptides
Mouse Protein Tnfrsf14, is a type I transmembrane protein belonging to the TNF receptor superfamily. It is tumour necrosis factor receptor superfamily member 14 and expressed on the surface of T cells during the resting state. Interaction of HVEM with TNF family member LIGHT co-stimulates T cells and promotes inflammation. HVEM also triggers inhibitory signalling cascade in effector T (Teff) cells and regulatory T cells (Tregs) as a ligand of B and T lymphocyte attenuator. Tnfrsf14 is detected in peripheral blood T cells, B cells, monocytes and in various tissues enriched in lymphoid cells. It has demonstrated that HVEM Ig is able to exert a significant antiviral effect against HSV-1 infection in vivo.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité