Rat Recombinant VEGFA (from P. pichia)
Fournisseur: ProSci Inc.
92-494
PRSI92-494EA 761 EUR InStock
PRSI92-494
Rat Recombinant VEGFA (from P. pichia)
Protéines et Peptides
Vascular endothelial growth factor (VEGF/VEGF-A ) is originally known as vascular permeability factor (VPF). It belongs to the PDGF family with a cysteine-knot structure comprised of eight conserved cysteine residues, and reckoned as a potent mediator in the process of angiogenesis and vasculogenesis in either fetus or adult. VEGF is particularly expressed in supraoptic , paraventricular nuclei and the choroid plexus of the pituitary, and abundant in the corpus luteum of the ovary and in kidney glomeruli. The rat VEGF protein contains a putative 20 amino acids (aa) signal peptide, and alternative splicing of rat VEGF gene produces isoforms of 120, 144, 164 and 188 aa. Rat VEGF164 respectively displays 97% and 88% aa identity with that regions of mouse and human VEGF. VEGF can bind to the FLT1/VEGFR1 and KDR/VEGFR2 receptors, heparan sulfate and heparin, and play important roles in inducing endothelial cell proliferation, promoting cell migration, inhibiting apoptosis and inducing permeabilisation of blood vessels.
  • This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité