Mouse recombinant PDL2
Fournisseur: ProSci Inc.
92-497
PRSI92-497EA 365 EUR InStock
PRSI92-497
Mouse recombinant PDL2
Protéines et Peptides
Programmed cell death 1 ligand 2 (PD-L2), also known as butyrophilin B7-DC or PDCD1 ligand 2, belongs to the member of B7 family which can regulate the activation and tolerance of T cells. PD-L2 is one ligand for Programmed cell death 1(PD-1), and the other is PD-L1. These two ligands shares 34% aa sequence identity. Mouse PD-L2 gene encodes a 273 amino acids (aa) protein with a putative 19 aa signal peptide, a 201 aa extracellular region , a 21 aa transmembrane domain and a 32 aa cytoplasmic region. The mouse PD-L2 gene is highly expressed in heart, placenta, pancreas, lung and liver while expressed weakly in spleen, lymph nodes and thymus. Besides, the expression of PD-L2 gene can be induced on dendritic cells grown from peripheral blood mononuclear cells under CSF2 and IL4/interleukin-4 treatment, and up-regulated by IFNG/IFN-gamma stimulation in monocytes. PD-L2 usually functions in a PDCD1-independent manner and is involved in regulating costimulatory signal which is essential for T-cell proliferation and IFNG production. Recent studies demonstrate that the expression of PD-L2 on the tumour cells promotes CD8 T cell–mediated rejection of tumour cells, at both the induction and effector phase of antitumour immunity. Moreover, PD-L2 binds to PD-1 cells and enhances T cell killing in a PD-1–independent mechanism.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité