Vibrio cholerae recombinant toxin B (from Escherichia coli)
Fournisseur: ProSci Inc.
92-498
PRSI92-498EA 760 EUR InStock
PRSI92-498
Vibrio cholerae recombinant toxin B (from Escherichia coli)
Protéines et Peptides
Cholera toxin is protein complex secreted by the bacterium Vibrio cholerae. It is responsible for the massive, watery diarrhea characteristic of cholera infection. Cholera enterotoxin subunit B (CTXB) pentameric ring directs the A subunit to its target by binding to the GM1 gangliosides present on the surface of the intestinal epithelial cells. It can bind five GM1 gangliosides. It has no toxic activity by itself.
  • This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité