Human recombinant sulphotransferase 1C4 (from E. coli)
Fournisseur: ProSci Inc.
92-500
PRSI92-500EA 584 EUR InStock
PRSI92-500
Human recombinant sulphotransferase 1C4 (from E. coli)
Enzymes
Human Sulfotransferase (SULT1C4) is an enzyme that in humans is encoded by the SULT1C4 gene, belongs to the sulfotransferase 1 family. SULT1C4 is expressed at high levels in fetal lung and kidney and at low levels in fetal heart, adult kidney, ovary and spinal chord. Sulfotransferase utilises 3'-phospho-5'-adenylyl sulfate (PAPS) as sulfonate donor to catalyse the sulfate conjugation of drugs, xenobiotic compounds, hormones, and neurotransmitters. It shows activity towards p-nitrophenol and N-hydroxy-2-acetylamino-fluorene (N-OH-2AAF). SULT1C4 plays an important role incatalyzing the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité