Human recombinant Biliverdin reductase A (from E. coli)
Fournisseur: ProSci Inc.
92-501
PRSI92-501EA 584 EUR InStock
PRSI92-501
Human recombinant Biliverdin reductase A (from E. coli)
Enzymes
Human Biliverdin reductase A (BLVRA) is belonged to the Gfo/Idh/MocA family and Biliverdin reductase subfamily. BLVRA is an enzyme that in humans is encoded by the BLVRA gene. BLVRA plays an important role in reducing the gamma-methene bridge of the open tetrapyrrole, biliverdin IX alpha, to bilirubin with the concomitant oxidation of a NADH or NADPH cofactor. BLVRA acts on biliverdin by reducing its double-bond between the pyrrole rings into a single-bond. It accomplishes this using NADPH + H+ as an electron donor, forming bilirubin and NADP+ as products.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité