Human Recombinant NTAQ1 (from E. coli)
Fournisseur: ProSci Inc.
92-506
PRSI92-506EA 654 EUR InStock
PRSI92-506
Human Recombinant NTAQ1 (from E. coli)
Protéines et Peptides
Human protein N-terminal glutamine amidohydrolase (WDYHV1) is an enzyme that in humans is encoded by the WDYHV1 gene, belongs to the NTAQ1 family.WDYHV1 mediates the side-chain deamidation of N-terminal glutamine residues to glutamate, which is an important step in N-end rule pathway of protein degradation. Conversion of the resulting N-terminal glutamine to glutamate renders the protein susceptible to arginylation, polyubiquitination and degradation as specified by the N-end rule. However,it does not act on substrates with internal or C-terminal glutamine andnon-glutamine residues in any position. With the exception of proline, all tested second-position residues on substrate peptides do not greatly influence the activity. In contrast, a proline at position 2, virtually abolishes deamidation of N-terminal glutamine.
Fusion-Tag: N-GST tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité