Mouse recombinant IL21 (from E. coli)
Fournisseur: ProSci Inc.
92-507
PRSI92-507EA 852 EUR InStock
PRSI92-507
Mouse recombinant IL21 (from E. coli)
Protéines et Peptides
Interleukin-21 also known as IL-21 is a protein that in mouse is encoded by the IL21 gene, belongs to the IL-15/IL-21 family. Interleukin-21 is a cytokine that has potent regulatory effects on cells of the immune system, including natural killer (NK) cells and cytotoxic T cells that can destroy virally infected or cancerous cells. This cytokine induces cell division/proliferation in its target cells.
  • This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité