Human recombinant FGF8F (from E. coli)
Fournisseur: ProSci Inc.
92-509
PRSI92-509EA 727 EUR InStock
PRSI92-509
Human recombinant FGF8F (from E. coli)
Protéines et Peptides
Fibroblast growth factor 8 (FGF­8) is a member of the fibroblast growth factor family. It is discovered as a growth factor essential for the androgen-­dependent growth of mouse mammary carcinoma cells. Mouse FGF­8b shares 100% aa identity with human FGF­8b. FGF­8 is widely expressed during embryogenesis, and mediates epithelial­-mesenchymal transitions. It plays an important role in the regulation of embryonic development, cell proliferation, cell differentiation and cell migration. It is required for normal brain, eye, ear, limb development during embryogenesis and normal development of the gonadotropin-releasing hormone (GnRH) neuronal system.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité