Mouse recombinant IL4 (from E. coli)
Fournisseur: ProSci Inc.
92-512
PRSI92-512EA 852 EUR InStock
PRSI92-512
Mouse recombinant IL4 (from E. coli)
Protéines et Peptides
Mouse Interleukin-4(IL-4) is a monomeric, Th2 cytokine that shows pleiotropic effects during immune responses. It is a glycosylated polypeptide that contains three intrachain disulfide bridges and adopts a bundled four alpha­helix structure. IL­4 exerts its effects through two receptor complexes, Participates in at least several B-cell activation processes as well as of other cell types. IL­4 is primarily expressed by Th2­biased CD4+T cells, mast cells, basophils, and eosinophils. It promotes cell proliferation, survival, and immunoglobulin class switch to IgG1 and IgE in mouse B cells, acquisition of the Th2 phenotype by naïve CD4+T cells, priming and chemotaxis of mast cells, eosinophils, and basophils, and the proliferation and activation of epithelial cells. IL­4 plays a dominant role in the development of allergic inflammation and asthma. It also regulates the expression of the low affinity Fc receptor for IgE (CD23) on both lymphocytes and monocytes.
  • This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité