Human recombinant Thymic stromal lymphopoietin (from E. coli)
Fournisseur: ProSci Inc.
92-513
PRSI92-513EA 852 EUR InStock
PRSI92-513
Human recombinant Thymic stromal lymphopoietin (from E. coli)
Protéines et Peptides
Thymic stromal lymphopoietin (TSLP) is a novel member of the hemopoietic cytokine family that promotes the development of B cells and shares overlapping activity with IL-7. The human TSLP protein comprises a 28 amino acids (aa) signal sequence and 131 aa mature region. Human TSLP has two isoforms lfTSLP (159 aa) and sfTSLP (63 aa) produced by alternative splicing . lfTSLP (159aa) is expressed in a number of tissues including heart, liver and prostate, and sfTSLP (63aa) is predominantly expressed in keratinocytes of oral mucosa, skin and in salivary glands. In aa sequence level, Human TSLP displays about 43% identity with mouse TSLP. Thymic stromal lymphopoietin (TSLP) is a cytokine that functions mainly on myeloid cells; it induces the release of T cell-attracting chemokines from monocytes and enhances the maturation of CD11c(+) dendritic cells . TSLP has proliferative effects on the myeloid cell line and may initiate asthma or atopic dermatitis responses by directly activating mast cells . TSLP signals cells via the interleukin-7 receptor- alpha chain (IL-7R alpha), shared with IL-7, together with the TSLP receptor (TSLPR) subunit. Recent studies indicate that TSLP and its receptor are novel therapeutic targets for rheumatoid arthritis , for increased intraarticular TSLP concentrations in patients has caused chemotaxis and activation of arthritogenic T cells.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité