Human recombinant Neutrophil cytosol factor 1 (from E. coli)
Fournisseur: ProSci Inc.
92-514
PRSI92-514EA 760 EUR InStock
PRSI92-514
Human recombinant Neutrophil cytosol factor 1 (from E. coli)
Protéines et Peptides
Neutrophil cytosol factor 1( NCF1) is a 47 kDa cytosolic subunit of neutrophil NADPH oxidase. This oxidase is characterised as a multicomponent enzyme which is activated to produce superoxide anion. NCF2, NCF1, and a membrane bound cytochrome b558 are required for the activation of the latent NADPH oxidase. The human NCF1 gene encodes a 390 amino acids protein without a signal peptide. The NCF1 gene interacts with other subunits of nicotinamide adenine dinucleotide phosphate-oxidase (NADPH) and plays an important role in innate immunity, producing reactive oxygen species and reducing the severity and duration of parasitic infection and autoimmune disease. NCF1 also has a role in T cell activation.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité