Human recombinant Apolipoprotein A-I (from E. coli)
Fournisseur: ProSci Inc.
92-516
PRSI92-516EA 518 EUR InStock
PRSI92-516
Human recombinant Apolipoprotein A-I (from E. coli)
Protéines et Peptides
Apolipoprotein A1 (APOA1) is a secreted protein which belongs to the Apolipoprotein A1/A4/E family. APOA1 is the major protein component of high density lipoprotein (HDL) in plasma. APOA1 plays a critical role in various biological processes, such as Cholesterol metabolism, Lipid metabolism and transport, Steroid metabolism. APOA1 promotes cholesterol efflux from tissues to the liver and thus helps to clear cholesterol from arteries. Defects in this gene resulted in HDL deficiencies, including Tangier disease (TGD), systemic non-neuropathic amyloidosis, premature coronary artery disease, hepatosplenomegaly and progressive muscle wasting and weakness. In addition, ApoA-I is implicated in the anti-endotoxin function of HDL via interaction with lipopolysaccharide or endotoxin.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité