Mouse recombinant IL1RA (from E. coli)
Fournisseur: ProSci Inc.
92-517
PRSI92-517EA 852 EUR InStock
PRSI92-517
Mouse recombinant IL1RA (from E. coli)
Protéines et Peptides
Interleukin-1 receptor antagonist protein (Il1rn), also known as IL-1ra, IRAP or IL1 inhibitor, is a member of the interleukin 1 cytokine family. This protein inhibits the activities of interleukin 1 alpha (IL1A) and interleukin 1 beta (IL1B), and modulates a variety of interleukin 1 related immune and inflammatory responses. The mouse Il1rn gene encodes a 178 amino acids (aa) protein with a 26 aa signal peptide. Mouse Il1rn protein shares 26% and 19% identity with its homologues IL-1 beta and IL-1 alpha, respectively. Il1rn can Inhibits the activity of interleukin-1 by binding to receptor IL1R1 and preventing its association with the coreceptor IL1RAP for signalling, but has no interleukin-1 like activity. Recently, an recombinant human Il1rn protein is used in the treatment of rheumatoid arthritis, an autoimmune disease in which IL-1 plays a key role.
  • This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité