Mouse recombinant NGF beta (from E. coli)
Fournisseur: ProSci Inc.
92-518
PRSI92-518EA 469 EUR InStock
PRSI92-518
Mouse recombinant NGF beta (from E. coli)
Protéines et Peptides
Mouse beta -NGF is a neurotrophic factor structurally related to BDNF, NT-3 and NT-4. These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures. beta -NGF is a potent neurotrophic factor that signals through its receptor beta -NGFR, and plays a crucial role in the development and preservation of the sensory and sympathetic nervous systems. beta -NGF also acts as a growth and differentiation factor for B lymphocytes, and enhances B-cell survival.
  • This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité