Mouse recombinant alpha-Synuclein (from E. coli)
Fournisseur: ProSci Inc.
92-520
PRSI92-520EA 844 EUR InStock
PRSI92-520
Mouse recombinant alpha-Synuclein (from E. coli)
Protéines et Peptides
Alpha-synuclein (Snca) belongs to a family of proteins including a-, b-, and g-synucleins. Alpha-synuclein has been found to be implicated in the pathophysiology of many neurodegenerative diseases, including Parkinson's disease (PD) and Alzheimer's disease. Manyneurodegenerative diseases has shown that alpha-synuclein accumulates in dystrophic neurites and in Lewy bodies. The function of alpha-synuclein is closely correlated with its three-dimensional structure, especially for proteins important in the pathogenesis of neurodegenerative diseases. Alpha-synuclein is a dynamic molecule whose secondary structure depends on the environment. For example, it has an unfolded random coil structure in aqueous solution, forms a-helical structure upon binding to acidic phospholipid vesicles, and forms insoluble fibrils with a high b-sheet content that resemble the filaments found in Lewy bodies. Also, alpha-synuclein was known to associate with 14-3-3 proteins including protein kinase C, BAD, and extracellular regulated kinase, and overexpression of alpha-synuclein could contribute to cell death in neurodegenerative diseases.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité