Mouse recombinant IL2 (from E. coli)
Fournisseur: ProSci Inc.
92-521
PRSI92-521EA 588 EUR InStock
PRSI92-521
Mouse recombinant IL2 (from E. coli)
Protéines et Peptides
Interleukin 2 (IL 2), also termed T-cell growth factor, is a member of the cytokine family which includes IL-4, IL-7, IL-9, IL-15 and IL-21. Each member of this family has a four alpha helix bundle. IL-2 signals through the IL-2 receptor, a complex consisting of tree subunits, termed alpha, beta and gamma. The IL-2 R gamma is shared by cytokine receptors of all members of cytokine family. Mature mouse IL­2 shares 56% and 73% aa sequence identity with human and rat IL­2, respectively. IL-2 is produced by CD4+ T cell, CD8+ T cells, gamma delta T cells, B cells, dendritic cells and eosinophils, and plays a vital role in key function of the immune system, tolerance and immunity, primarily via its potent stimulatory activity for T cells.Thus, IL­2 may be a key cytokine in the natural suppression of autoimmunity.
  • This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité