Mouse recombinant IL36 beta (from E. coli)
Fournisseur: ProSci Inc.
92-522
PRSI92-522EA 654 EUR InStock
PRSI92-522
Mouse recombinant IL36 beta (from E. coli)
Protéines et Peptides
Mouse Interleukin 36 beta (IL-36B)is a member of the IL-1 family of proteins. It is a cytokine that binds to and signals through the IL1RL2/IL-36R receptor which in turn activates NF-kappa-B and MAPK signalling pathways in target cells linked to a pro-inflammatory response. IL-36B is synthesised in several cells including resting and activated monocytes, and B cells. The receptor for IL-36 beta is thought to be a combination of IL-1 Rrp2 and IL-1 RAcP. Interleukin 36 beta is one part of the IL-36 signalling system that is thought to be present in epithelial barriers and to take part in local inflammatory response; similar to the IL-1 system with which it shares the coreceptor IL1RAP. Interleukin 36 beta are involved in a number of fundamental biological processes such as stimulating production of interleukin-6 and interleukin-8 in synovial fibrobasts, articular chondrocytes and mature adipocytes, inducing expression of a number of antimicrobial peptides including beta-defensin 4 and beta-defensin 103 as well as a number of matrix metalloproteases , inducing the production of proinflammatory cytokines in bone marrow-derived dendritic cells (BMDCs), including IL-12, Il-1 beta, IL-6, TNF-alpha and IL-23, and activating p38 MAPK phosphorylation in BMDCs.Moreover, interleukin 36 beta may be involved in skin inflammatory response by acting on keratinocytes, dendritic cells, and indirectly on T cells to drive tissue infiltration, cell maturation and cell proliferation. It plays an important role in dendritic cell maturation by stimulating the surface expression of CD80, CD86 and MHC class II and inducing the production of IFN-gamma, IL-4 and IL-17 by T helper 1 (Th1) cells, cultured CD4+ T cells and splenocytes.
  • This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité