Mouse recombinant LMIR1
Fournisseur: ProSci Inc.
92-527
PRSI92-527EA 528 EUR InStock
PRSI92-527
Mouse recombinant LMIR1
Protéines et Peptides
LMIR1, also termed CD300a, is a type I transmembrane glycoprotein with a single IgV-like extracellular domain and an extended membrane proximal region that links the immunoglobulin (Ig) and transmembrane domains and belongs to the immunoglobulin superfamily. The intracellular domain of LMIR1 contains several immunoreceptor tyrosine-based inhibition motifs (ITIMs). When cross-linked, it will be tyrosine phosphorylated and capable of recruiting tyrosine phosphatases (SHP-1, SHP-2) and inositol polyphosphate 5-phosphatase, SHIP. LMIR1 will regulate mast cell-mediated inflammatory responses. LMIR1 is broadly expressed on myeloid and lymphoid cells, and its expression is differentially regulated depending on the cell type.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité