Mouse recombinant TSLP receptor
Fournisseur: ProSci Inc.
92-533
PRSI92-533EA 678 EUR InStock
PRSI92-533
Mouse recombinant TSLP receptor
Protéines et Peptides
The cytokine thymic stromal lymphopoietin receptor (TSLPR) is consisting of a common gamma receptor–like chain (TSLPR- gamma ) and a common interleukin 7 (IL-7) R alpha chain that belongs to the type 1 cytokine receptor family. Transfection of TSLPR cDNA result in only low affinity binding, while cotransfection of the IL-7R alpha chain cDNA shows high affinity binding. TSLP and TSLPR play a critical role in the initiation of allergic diseases in mice. The TSLP R cDNA encodes a transmembrane receptor containing 370 amino acids (aa) with two potential N-linked glycosylation sites and a cytoplasmic domain of 104 aa including a single tyrosine residue. TSLPR can mediate signalling of the signal transducer and activator of transcription 5 (Stat5) by TSLP. TSLP R is broadly expressed in the immune and hematopoietic cells, particularly in hematopoietic progenitors and myeloid cells.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité