Mouse recombinant pentraxin 2
Fournisseur: ProSci Inc.
92-536
PRSI92-536EA 844 EUR InStock
PRSI92-536
Mouse recombinant pentraxin 2
Protéines et Peptides
Pentraxin 2 (PTX2), also known as Serum amyloid P (SAP), is a highly conserved, naturally circulating plasma protein and a soluble pattern recognition receptor of the innate immune system. The unique binding activities indicated that it may play an important role in the removal of damaged tissue. PTX2 belongs to the pentraxin family, is universally present in amyloid deposits. Mouse with targeted deletion of the PTX2 gene shows retarded and reduced induction of experimental reactive systemic (AA type) amyloidosis confirmed that it does indeed contribute to pathogenesis of amyloidosis and is a valid therapeutic target. In recent discovery, PTX2 can be used as a powerful antifibrotic agent to regulate certain monocyte differentiation states.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité