Mouse recombinant IFN gamma R1
Fournisseur: ProSci Inc.
92-539
PRSI92-539EA 409 EUR InStock
PRSI92-539
Mouse recombinant IFN gamma R1
Protéines et Peptides
The tetrameric receptor complex for IFN gamma consists of two subunits, IFNGR1 (IFN gamma R alpha) and IFNGR2 (IFN gamma R beta ), through which the dimeric IFN- gamma exerts its biological functions, including antiviral, antiproliferation and immune-modulatory activity in mammals. Both IFNGR1 and IFNGR2 are single transmembrane proteins belonging to the class II cytokine family. FNGR1, widely expressed in most host cells, is essential for IFN gamma binding, receptor trafficking, and signal transduction. IFNGR1 possesses an intracellular Janus tyrosine kinase (JAK) 1 binding site, a signal transducer and activator of transcription 1 (STAT1) binding site. The resulting STAT1 homodimers translocate from the cytoplasm to the nucleus and bind to the interferon-gamma activated sequence (GAS) promoter to induce expression of downstream interferon stimulated genes (ISGs).
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité