Mouse recombinant carboxylesterase 2E
Fournisseur: ProSci Inc.
92-540
PRSI92-540EA 852 EUR InStock
PRSI92-540
Mouse recombinant carboxylesterase 2E
Enzymes
Carboxylesterase 5 (CES5), also called cauxin or CES7, is a member of carboxylesterases family which plays an important role in the hydrolysis of ester and amide bonds. Carboxylesterase is a type of enzyme that capable of hydrolysing a variety of carboxylic acid esters and it's widely distributed in cells especially in mammalian liver. CES5 is with broad substrate specificity ranging from small molecule esters to longchain fatty acid esters and thioesters. It has been previously reported CES5 was in high concentrations in the urine (cauxin) of adult male cats, and within a protein complex of mammalian male epididymal fluids. Roles for CES5 may include regulating urinary levels of male cat pheromones, catalysing lipid transfer reactions within mammalian male reproductive fluids, and protecting neural tissue from drugs and xenobiotics.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité