Mouse recombinant cathepsin H
Fournisseur: ProSci Inc.
92-542
PRSI92-542EA 852 EUR InStock
PRSI92-542
Mouse recombinant cathepsin H
Protéines et Peptides
Cathepsin H (CTSH), which can act both as an aminopeptidase and as an endopeptidase, is a lysosomal cysteine protease of the papain family. CTSH is composed of a dimer of disulfide-linked heavy and light chains, both produced from a single protein precursor. CTSH is associated with various pathological conditions like human fibrous meningioma, colorectal cancer, arthritis, human prostate tumour and lung cancer. CTSH is associated with cancer progression because of their ability to degrade extracellular matrices facilitating invasion, angiogenesis and metastasis as is evident from numerous clinical reports and experimental models. The expression of CTSH is significantly increased in disease states such as in prostate tumours, sera of asthmatic patients, and mucosa of colorectal cancer patients.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité