Human recombinant ULBP-1 (from cells)
Fournisseur: ProSci Inc.
92-545
PRSI92-545EA 469 EUR InStock
PRSI92-545
Human recombinant ULBP-1 (from cells)
Protéines et Peptides
NKG2D ligand 1, also called ULBP1, is a member of UL16-binding protein (ULBP) family which has also been termed the retinoic acid early transcript 1 (RAET1) family. Unlike the classical MHC class I molecules and the MIC molecules possess alpha1, alpha2 and alpha3 domains, ULBP/RAET1 family members lack alpha3 domain. ULBP1 is recognised by the activating receptor NKG2D on the surface of cytotoxic natural killer (NK) and T cells, and then promotes the lysis of cells expressing ULBP1 which is important for the immune surveillance. ULBP1 and several other family members, ULBP2 and ULBP5, own the ability to bind the human cytomegalovirus (CMV) UL16 glycoprotein. The human CMV glycoprotein UL16 binds to intracellular ULBP1 and so inhibits its expression at the cell surface, which reduces the susceptibility of the virus-infected cell to cytotoxic destruction by NK cells. The expression of ULBP1 has been found on some tumour cells and is implicated in tumour surveillance.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité