Human recombinant Ephrin type-A Receptor 4 (from cells)
Fournisseur: ProSci Inc.
92-546
PRSI92-546EA 419 EUR InStock
PRSI92-546
Human recombinant Ephrin type-A Receptor 4 (from cells)
Protéines et Peptides
Ephrin type-A receptor 4(EPHA4) belongs to the protein kinase superfamily and Ephrin receptor subfamily. EPHA4 contains 1 Eph LBD domain, 2 fibronectin type-III domains, 1 protein kinase domain and 1 SAM domain. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité