Mouse recombinant OX40
Fournisseur: ProSci Inc.
92-549
PRSI92-549EA 469 EUR InStock
PRSI92-549
Mouse recombinant OX40
Protéines et Peptides
OX40, also termed CD134 and TNFRSF4, is a T cell co-stimulatory molecule of the TNF receptor superfamily which plays a key role in the survival and homeostasis of effector and memory T cells. OX40 is expressed on CD4+ and CD8+ T cells upon engagement of the TCR by antigen presenting cells along with co-stimulation by CD40-CD40 Ligand and CD28-B7. The interaction between OX40 and OX40 ligand (OX40 L) will occur when activated T cells bind to professional antigen-presenting cells (APCs). The T-cell functions, including cytokine production, expansion, and survival, are then enhanced by the OX40 costimulatory signals. OX40 signals are critical for controllling the function and differentiation of Foxp3+ regulatory T cells. OX40-OX40L interaction regulates T-cell tolerance, peripheral T-cell homeostasis, and T-cell-mediated inflammatory diseases.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité