Human recombinant RANK (from cells)
Fournisseur: ProSci Inc.
92-559
PRSI92-559EA 435 EUR InStock
PRSI92-559
Human recombinant RANK (from cells)
Protéines et Peptides
Receptor Activator of Nuclear Factor κ B (RANK), also known as CD265, TRANCE Receptor or TNFRSF11A, is member of the tumour necrosis factor receptor (TNFR) molecular superfamily. RANK is the receptor for RANK-Ligand (RANKL) and part of the RANK/RANKL/OPG signallling pathway that regulates osteoclast differentiation and activation. It plays a vital role in bone remodellling and repair, immune cell function, lymph node development, thermal regulation, and mammary gland development. RANK is constitutively expressed in skeletal muscle, thymus, liver, colon, small intestine, adrenal gland, osteoclast, mammary gland epithelial cells, prostate, vascular cell, and pancreas.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité