Mouse recombinant CD5
Fournisseur: ProSci Inc.
92-560
PRSI92-560EA 588 EUR InStock
PRSI92-560
Mouse recombinant CD5
Protéines et Peptides
CD5 is a transmembrane glycoprotein of the conserved scavenger receptor cysteine-rich (SRCR) superfamily and expressed on thymocytes, peripheral T cells and a subset of B cells (B1-a). Moreover, CD5 also was found expressed in small lymphocytic lymphoma, hairy cell leukaemia and mantle cell lymphoma cells. The long cytoplasmic tail of CD5 has no intrinsic enzymatic activity, but contains four tyrosine phosphorylation sites, including an immunoreceptor tyrosine-based (ITAM)-like motif (pseudo-ITAM) and an immunoreceptor tyrosine-based inhibitory (ITIM)-like motif (pseudo-ITIM), as well as multiple potential serine and threonine phosphorylation sites. It physically associates with the T cell antigen receptor (TCR) and B cell antigen receptor (BCR), where it negatively modulates the activation and differentiation signals transduced by these receptors. CD5 also plays an important role in protection from activation-induced cell death and in the recognition of pathogen associated molecular patterns (PAMPS) present on fungal surfaces.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité