Mouse recombinant DAN
Fournisseur: ProSci Inc.
92-561
PRSI92-561EA 588 EUR InStock
PRSI92-561
Mouse recombinant DAN
Protéines et Peptides
Differential screening-selected gene aberrative in neuroblastoma (DAN) is a member of the DAN family of secreted glycoproteins. DAN family antagonists are Characterised by a DAN domain that contains a cystine knot motif which is essential for binding to BMP ligands. Members of this family include DAN, gremlin, protein related to DAN and cerberus (PRDC), cerberus, sclerostin (SOST) and uterine sensitisation-associated gene 1 protein, and control diverse processes in growth, development and the cell cycle. It has also been reported that DAN family plays crucial role in early mouse embryo development by inhibiting the action of bone morphogenic proteins and modulating the action of transforming growth factor- beta superfamily members. DAN is Synthesised by small-to intermediate-sised DRG neurons and transported to the sensory nerve terminals in the skin or to the sensory nerve terminals in the dorsal horn. It has been reported that DAN is ubiquitously expressed in adult rat and human tissues. Morphological studies have revealed that, in adult rat, DAN mRNA is expressed ubiquitously in lung and brain, but not in liver.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité