Mouse recombinant carbonic anhydrase 12
Fournisseur: ProSci Inc.
92-563
PRSI92-563EA 852 EUR InStock
PRSI92-563
Mouse recombinant carbonic anhydrase 12
Enzymes
Carbonic Anhydrase (CA) XII, also known as Car12 and CA12, is an extracellular enzyme involved in the regulation of the microenvironment acidity and tumour malignant phenotype, was originally identified as a protein overexpressed in some types of cancers. It has showed that CA XII is induced by hypoxia and oestrogen and expressed at high levels on various types of cancer. The enzyme is directly involved in tumour progression, and its inhibition has an anti-tumour effect. Apart from its role in carcinogenesis, the enzyme contributes to various other diseases like glaucoma and arteriosclerotic plaques, among others. CA XII is therefore regarded as promising target for specific therapies, and may be used as a novel prognostic marker in combination with histologic grade of the tumours.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité