Mouse recombinant ALK1
Fournisseur: ProSci Inc.
92-565
PRSI92-565EA 343 EUR InStock
PRSI92-565
Mouse recombinant ALK1
Protéines et Peptides
Activin Receptor-Like Kinase 1 (ALK-1) is a type I cell-surface receptor for the TGF- beta superfamily of ligands, which mediates signallling of BMP9 (bone morphogenetic protein) and BMP10. ALK1 signallling is necessary for angiogenesis during embryogenesis, wound healling, and tumour growth. ALK-1 has a high degree of similarity in serine-threonine kinase subdomains, a glycine and serine rich region preceding the kinase-domain, and a C-terminal tail with other activin receptor-like kinase proteins. ALK-1 is mainly expressed in endothelial cells regulating proliferation and migration in vitro and angiogenesis in vivo. Mutations in ALK-1 as well as in endoglin are associated with hereditary haemorrhagic telangiectasia (HHT), suggesting ALK-1 plays a critical role for in the control of blood vessel development or repair.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité