Mouse recombinant ectodysplasin receptor
Fournisseur: ProSci Inc.
92-570
PRSI92-570EA 588 EUR InStock
PRSI92-570
Mouse recombinant ectodysplasin receptor
Protéines et Peptides
Ectodysplasin A receptor (EDAR) is a type I transmembrane protein of the TNF- alpha receptor superfamily which plays a key role in ectodermal differentiation. EDAR was encoded by the mouse down less gene and defective in human dominant and recessive forms of autosomal hypo hidrotic ectodermal dysplasia (EDA) syndrome. The extracellular domain of EDAR contains 14 cysteine residues, six of which approximate the TNFRSF cysteine-rich region, the cytoplasmic domain contains a region with homology to the death domains found in other TNFRSF members. EDAR has been suggested to be an early and important promoter of placode development in all ectodermal organs, such as uch as hair follicles, teeth and sweat glands. EDA-A1, the A1 isoform of EDA, is the EDAR ligand. EDA and EDA are implicated in appendage development by the cloning of a gene underlying hypo hidrotic ectodermal dysplasia (HED) in mouse and human. HED is Characterised by agenesis or malformation of ectoderm-derived appendages, such as teeth, sweat glands and hair follicles, while the skin itself develops normally.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité