Mouse recombinant B7-2
Fournisseur: ProSci Inc.
92-573
PRSI92-573EA 306 EUR InStock
PRSI92-573
Mouse recombinant B7-2
Protéines et Peptides
The protein is the receptor that involved in the costimulatory signal essential for T-lymphocyte proliferation and interleukin-2 production, by binding CD28 or CTLA-4. It may play a critical role in the early events of T-cell activation and stimulation of naive T-cells, such as deciding between immunity and anergy that is made by T-cells within 24 hours after activation. Isoform 2 interferes with the formation of CD86 clusters, and thus acts as a negative regulator of T-cell activation. The protein interacts with MARCH8, human herpesvirus 8 MIR2 protein, adenovirus subgroup B fibre proteins and acts as a receptor for these viruses. It is expressed by activated B-lymphocytes and monocytes and promoted by MARCH8 and results in endocytosis and lysosomal degradation. It contains 1 Ig-like C2-type(immunoglobulin-like) domain and 1 Ig-like V-type (immunoglobulin-like) domain.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité