Human recombinant syntaxin 7 (from cells)
Fournisseur: ProSci Inc.
92-579
PRSI92-579EA 588 EUR InStock
PRSI92-579
Human recombinant syntaxin 7 (from cells)
Protéines et Peptides
Syntaxin 7 (STX7) is a member of a family of membrane-trafficking proteins named as soluble N-ethylmaleimide-sensitive factor attachment protein receptors (SNAREs), and mediates the endocytic trafficking from early endosomes to late endosomes and lysosomes. Syntaxin-7 has been shown to be present in both late endosomes and lysosomes, and to be required for both homotypic late endosome fusion and heterotypic fusion with lysosomes. STX7 has been shown to interact with STX8, VPS18, Vesicle-associated membrane protein 8 and VPS11.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité