Mouse recombinant B7-1
Fournisseur: ProSci Inc.
92-581
PRSI92-581EA 343 EUR InStock
PRSI92-581
Mouse recombinant B7-1
Protéines et Peptides
Cluster of Differentiation 80, also called B7-1, is a member of cell surface immunoglobulin superfamily which plays key, yet distinct roles in the activation of T cells. It is the ligand for two different proteins on the T cell surface: CD28 and CTLA-4. Studies have shown that CTLA-4 binds mostly to CD80. The structure presents two extracellular domains: a membrane distal variable-like domain (IgV) and a membrane proximal Ig constant-like domain (IgC) along with an intracellular domain. Both IgV and IgC consist of anti-parallel beta sandwiches joined by a short linker region. CD80 is mostly expressed on the surface of antigen-presenting cells including activated B cells, macrophages and dendritic cells.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité