Mouse recombinant ST2
Fournisseur: ProSci Inc.
92-583
PRSI92-583EA 588 EUR InStock
PRSI92-583
Mouse recombinant ST2
Protéines et Peptides
ST2, also called IL-1 R4, is an Interleukin-1 receptor family glycoprotein that plays a role in Th2 immune responses. ST2 is expressed on the surface of mast cells, activated Th2 cells, macrophages, and cardiac myocytes. This receptor is very similar to the IL-1 receptor type I and the IL-18 receptor alpha chain in that ST2 also has three extracellular Ig domains and an intracellular Toll domain. ST2 binds IL-33, enhances inflammatory cytokines by activating nuclear factor-κB (NF-κB) and mitogen activated protein (MAP) kinases. ST2 exists as either a membrane bound form (ST2L) or as a soluble form (sST2). ST2L acts as a transmembrane signalling receptor for IL-33 by mediating the effect of IL-33 on the inflammatory process, while sST2 can suppress IL-33 activity.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité