Mouse recombinant serpin D1
Fournisseur: ProSci Inc.
92-592
PRSI92-592EA 852 EUR InStock
PRSI92-592
Mouse recombinant serpin D1
Protéines et Peptides
SerpinD1, also known as heparin cofactor II(HC-II), is a member of Serpin superfamily of the serine proteinase inhibitors. It is a single chain glycoprotein with a size of 66.5 kDa and is secreted from hepatocytes. HC-II acts as a thrombin inhibitor in the coagulation cascade, in a glycosaminoglycan-dependent pathway using the release of a sequestered hirudin-like N-terminal tail for interaction with thrombin. This serpin belongs to multiple member group V2 of vertebrate serpin classification. It has been suggested that HC-II is a predictor of decreased atherosclerosis in the elderly and protective against atherosclerosis in mice. HCII can used as a predictive biomarker and therapeutic target for atherosclerosis.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité