Mouse recombinant TGF beta Receptor II
Fournisseur: ProSci Inc.
92-598
PRSI92-598EA 461 EUR InStock
PRSI92-598
Mouse recombinant TGF beta Receptor II
Protéines et Peptides
Transforming growth factor- beta (TGF- beta ) is an essential regulator in the processes of development, cell proliferation, and extracellular matrix deposition. TGF- beta regulates cellular processes by binding to three high-affinity cell surface receptors: TGF- beta receptor type I (TGF- beta -RI), TGF- beta receptor type II (TGF- beta -RII), and TGF- beta beta receptor type III (TGF- beta -RIII). TGF- beta RII is consists of a C-terminal protein kinase domain and an N-terminal ectodomain and belongs to transforming growth factor-beta (TGF- beta ) receptor subfamily. TGF- beta RII has a protein kinase domain which can form a heterodimeric complex with another receptor protein and bind TGF-beta. This receptor/ligand complex phosphorylates protein will enter the nucleus and regulate the transcription of a subset of genes related to cell proliferation.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité