Mouse recombinant CREG1
Fournisseur: ProSci Inc.
92-606
PRSI92-606EA 654 EUR InStock
PRSI92-606
Mouse recombinant CREG1
Protéines et Peptides
Cellular repressor of E1A genes (CREG) is an evolutionarily conserved lysosomal protein, and an important new factor in regulating tissue homeostasis that has been shown to antagonise injury of tissues or cells. CREG contains three mannose 6-phosphate (M6P) markers, and depends on interactions with M6P receptors for efficient delivery to lysosomes, which is implicated in the regulation of lysosomal functions. This protein shares limited sequence similarity with E1A and binds both the general transcription factor TBP and the tumor suppressor pRb in vitro. CREG plays an important role in the control of cell growth and differentiation. It has been shown that CREG antagonises transcriptional and cellular transformation by the adenoviral E1A oncoprotein, induces differentiation while attenuating cellular proliferation, regulates the levels of the signaling kinases ERK1/2, and mediates glucocorticoid-induced proliferation of ileal epithelial cells. CREG is widely expressed in adult tissues, such as the brain, heart, lungs, liver, intestines and kidneys in mice, but is not markedly expressed in pluripotent embryonic stem cells.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité