Mouse recombinant NGAL
Fournisseur: ProSci Inc.
92-610
PRSI92-610EA 419 EUR InStock
PRSI92-610
Mouse recombinant NGAL
Protéines et Peptides
Lipocalin-2, also known as Neutrophil Gelatinase-Associated Lipocalin (NGAL), is a secretory protein of the lipocalin superfamily. Lipocalin-2 contains a signal peptide that enables it to be secreted and form complexes with matrix metalloproteinase-9 (MMP-9) through disulphide bonds. Similar to other lipocalin family members, Lipocalin-2 is involved in diverse cellular processes, including the transport of small hydrophobic molecules, protection of MMP-9 from proteolytic degradation, and cell signalling. Furthermore, Lipocalin-2 can tightly bind to bacterial siderophore through a cell surface receptor, possibly serving as a potent bacteriostatic agent by sequestering iron, regulating innate immunity and protecting kidney epithelial cells from ischemia–reperfusion injury. This protein is mainly expressed in neutrophils and in lower levels in the kidney, prostate, and epithelia of the respiratory and alimentary tracts. Recent evidence also suggests its role as a biomarker for renal injury and inflammation.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité