Carassiusauratus recombinant leptin (from P. pichia)
Fournisseur: ProSci Inc.
92-612
PRSI92-612EA 584 EUR InStock
PRSI92-612
Carassiusauratus recombinant leptin (from P. pichia)
Protéines et Peptides
Leptin is a hormone secreted from white adipocytes and plays important role in the regulation of food intake and energy balance. Leptin functions via signalling pathways involving OB-R in hypothalamus. In mammals, leptin is mainly produced by the adipose tissue and encodes body fat reserves, acting as a short-term satiety signal. In fish, the presence of a leptin-like peptide was first evidenced by immuno-cross-reactivity [14], and its existence was certainly demonstrated after the finding by synteny of a leptin sequence in the pufferfish.
Fusion-Tag: N-8 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité