Human recombinant Serpin B3 (from E. coli)
Fournisseur: ProSci Inc.
92-616
PRSI92-616EA 852 EUR InStock
PRSI92-616
Human recombinant Serpin B3 (from E. coli)
Protéines et Peptides
Serpin B3 is a protein that in humans is encoded by the SERPINB3 gene, belongs to the serpin family, Ov-serpin subfamily, May act as a papain-like cysteine protease inhibitor to modulate the host immune response against tumour cells. Also functions as an inhibitor of UV-induced apoptosis via suppression of the activity of c-Jun NH(2)-terminal kinase (JNK1).
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité