Human recombinant phosphoglucomutase-2 (from cells)
Fournisseur: ProSci Inc.
92-619
PRSI92-619EA 584 EUR InStock
PRSI92-619
Human recombinant phosphoglucomutase-2 (from cells)
Enzymes
Phosphoglucomutase-2 (PGM2) is a member of PGM family, which catalyses the inter-conversion of sugar phosphates and participates in anabolic and catabolic reactions. When cells are grown in glucose, PGM catalyses the conversion of glucose-6-phosphate to glucose-1-phosphate an important precursor required for the synthesis of UDP glucose and trehalose. PGM2 catalyses the conversion of the nucleoside breakdown products ribose-1-phosphate and deoxyribose-1-phosphate to the corresponding 5-phosphopentoses, and it may also catalyse the interconversion of glucose-1-phosphate and glucose-6-phosphate. But this protein has low glucose 1,6-bisphosphate synthase activity.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité