Human recombinant FSH (from Cells)
Fournisseur: ProSci Inc.
92-620
PRSI92-620EA 852 EUR InStock
PRSI92-620
Human recombinant FSH (from Cells)
Protéines et Peptides
Human Follicle-stimulating hormone (FSH) is a member of glycoprotein hormones subunit beta family, which also includes LH, chorionic gonadotropin (CG) and thyroid-stimulating hormone (TSH). FSH and its family members are heterodimers consisting of non-covalently linked alpha- and beta -subunits. They share an identical alpha subunit, and beta -subunits vary. FSH has a unique beta -subunit (FSH beta ), which confers its specific biologic activity and is responsible for interaction with the FSH-receptor which belongs to a subfamily of GPCRs called leucine-rich-repeat-containing GPCRs (LGRs). FSH is secreted from the pituitary gland and regulates reproduction in mammals. FSH stimulates Sertoli cell proliferation in testes and supports spermatogenesis in males, and induces the maturation of ovarian follicles in females.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité